Assay Details
| Gene Symbol: ALG3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943400.3 | XP_004943457.1 | 2 - 3 | 664 | 67 | |
| XM_004943402.3 | XP_004943459.1 | 3 - 4 | 456 | 67 | ||
| XM_015291555.2 | XP_015147041.1 | 2 - 3 | 664 | 67 | ||
| XM_422759.6 | XP_422759.4 | 3 - 4 | 440 | 67 | ||
More Information
Set Membership: |
|

ALG3
Genomic Map