Assay Details
| Gene Symbol: FAM162A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142894.1 | XP_024998662.1 | 2 - 3 | 197 | 73 | |
| XM_025142897.1 | XP_024998665.1 | 2 - 3 | 275 | 73 | ||
| GenBank mRNA | BX930601.2 | - | 2 - 3 | 299 | 73 | |
| BX935537.2 | - | 2 - 3 | 256 | 73 | ||
| CR406067.1 | - | 2 - 3 | 265 | 73 | ||
| CR407355.1 | - | 2 - 3 | 182 | 73 | ||
More Information
Set Membership: |
|

FAM162A
Genomic Map