Assay Details
| Gene Symbol: DAPK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015278951.2 | XP_015134437.1 | 2 - 3 | 218 | 97 | |
| GenBank mRNA | BX930723.2 | - | 1 - 2 | 107 | 97 | |
More Information
Set Membership: |
|

DAPK2
Genomic Map