Assay Details
| Gene Symbol: C18H17orf75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004946179.3 | XP_004946236.2 | 9 - 10 | 1225 | 138 | |
| XM_004946180.3 | XP_004946237.2 | 8 - 9 | 1155 | 138 | ||
| XM_415664.6 | XP_415664.3 | 9 - 10 | 1226 | 138 | ||
More Information
Set Membership: |
|

C18H17orf75
Genomic Map