Assay Details
| Gene Symbol: IST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015292719.2 | XP_015148205.1 | 4 - 5 | 408 | 72 | |
| XM_015292720.2 | XP_015148206.1 | 4 - 5 | 620 | 72 | ||
| XM_015292721.2 | XP_015148207.1 | 4 - 5 | 410 | 72 | ||
| XM_025154350.1 | XP_025010118.1 | 4 - 5 | 617 | 72 | ||
| XM_025154351.1 | XP_025010119.1 | 4 - 5 | 439 | 72 | ||
| XM_025154352.1 | XP_025010120.1 | 4 - 5 | 410 | 72 | ||
More Information
Set Membership: |
|

IST1
Genomic Map