Assay Details
| Gene Symbol: HEY1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282862.2 | XP_015138348.2 | 2 - 3 | 367 | 90 | |
| XM_025148013.1 | XP_025003781.1 | 2 - 3 | 371 | 90 | ||
More Information
Set Membership: |
|

HEY1
Genomic Map