Assay Details
| Gene Symbol: ABCA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004946232.3 | XP_004946289.1 | 34 - 35 | 4709 | 95 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ABCA8| Gene Symbol: ABCA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004946232.3 | XP_004946289.1 | 34 - 35 | 4709 | 95 | |
Set Membership: |
|