Assay Details
| Gene Symbol: TOMM34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417366.6 | XP_417366.3 | 4 - 5 | 547 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TOMM34| Gene Symbol: TOMM34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_417366.6 | XP_417366.3 | 4 - 5 | 547 | 94 | |
Set Membership: |
|