Assay Details
| Gene Symbol: IL1RAPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_425563.6 | XP_425563.3 | 10 - 11 | 2335 | 127 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IL1RAPL1| Gene Symbol: IL1RAPL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_425563.6 | XP_425563.3 | 10 - 11 | 2335 | 127 | |
Set Membership: |
|