Assay Details
| Gene Symbol: TMEM255B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277906.2 | XP_015133392.1 | 5 - 6 | 514 | 87 | |
| XM_015277907.2 | XP_015133393.1 | 6 - 7 | 605 | 87 | ||
| XM_025146697.1 | XP_025002465.1 | 6 - 7 | 764 | 87 | ||
| XM_025146698.1 | XP_025002466.1 | 6 - 7 | 674 | 87 | ||
| XM_025146699.1 | XP_025002467.1 | 6 - 7 | 764 | 87 | ||
| XM_025146700.1 | XP_025002468.1 | 6 - 7 | 764 | 87 | ||
| XM_025146701.1 | XP_025002469.1 | 6 - 7 | 629 | 87 | ||
| XM_025146702.1 | XP_025002470.1 | 6 - 7 | 764 | 87 | ||
| XM_025146703.1 | XP_025002471.1 | 5 - 6 | 539 | 87 | ||
| XM_025146704.1 | XP_025002472.1 | 5 - 6 | 521 | 87 | ||
| XM_025146706.1 | XP_025002474.1 | 6 - 7 | 695 | 87 | ||
| XM_025146707.1 | XP_025002475.1 | 7 - 8 | 813 | 87 | ||
| XM_025146708.1 | XP_025002476.1 | 6 - 7 | 599 | 87 | ||
| XM_025146709.1 | XP_025002477.1 | 6 - 7 | 661 | 87 | ||
| XM_025146710.1 | XP_025002478.1 | 4 - 5 | 476 | 87 | ||
| XM_025146711.1 | XP_025002479.1 | 6 - 7 | 629 | 87 | ||
| XM_025146712.1 | XP_025002480.1 | 5 - 6 | 484 | 87 | ||
| XM_025146713.1 | XP_025002481.1 | 4 - 5 | 431 | 87 | ||
More Information
Set Membership: |
|

TMEM255B
Genomic Map