Assay Details
| Gene Symbol: SULT4A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015291195.2 | XP_015146681.1 | 5 - 6 | 720 | 84 | |
| XM_015291200.2 | XP_015146686.1 | 5 - 6 | 665 | 84 | ||
| XM_015291212.2 | XP_015146698.1 | 5 - 6 | 720 | 84 | ||
| XM_025142261.1 | XP_024998029.1 | 5 - 6 | 488 | 84 | ||
More Information
Set Membership: |
|

SULT4A1
Genomic Map