Assay Details
| Gene Symbol: NEU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025153538.1 | XP_025009306.1 | 3 - 4 | 507 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_025153538.1 transcripts.
Set Membership: |
|

NEU2
Genomic Map