Assay Details
| Gene Symbol: POLR1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426271.6 | XP_426271.2 | 4 - 5 | 316 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

POLR1D| Gene Symbol: POLR1D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426271.6 | XP_426271.2 | 4 - 5 | 316 | 73 | |
Set Membership: |
|