Assay Details
| Gene Symbol: YDJC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001234390.5 | XP_001234391.1 | 7 | 3709 | 93 | |
| XM_025155736.1 | XP_025011504.1 | 6 | 4174 | 93 | ||
More Information
Set Membership: |
|

YDJC
Genomic Map