Assay Details
| Gene Symbol: LOC112529960 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144318.1 | XP_025000086.1 | 1 | 1282 | 103 | |
| Gene Symbol: MARCH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_418182.4 | XP_418182.2 | 5 | 2651 | 103 | |
More Information
Set Membership: |
|

LOC112529960
Genomic Map