Assay Details
| Gene Symbol: FBXO44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297211.2 | XP_015152697.1 | 5 - 6 | 716 | 153 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FBXO44| Gene Symbol: FBXO44 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297211.2 | XP_015152697.1 | 5 - 6 | 716 | 153 | |
Set Membership: |
|