Assay Details
| Gene Symbol: HTR1F | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004938334.3 | XP_004938391.1 | 5 | 5768 | 103 | |
| XM_004938336.3 | XP_004938393.1 | 4 | 5674 | 103 | ||
More Information
Set Membership: |
|

HTR1F
Genomic Map