Assay Details
| Gene Symbol: TIMP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232058.5 | XP_001232059.1 | 1 - 2 | 312 | 112 | |
| XM_025154764.1 | XP_025010532.1 | 2 - 3 | 270 | 112 | ||
| GenBank mRNA | BX931111.1 | - | 4 - 5 | 402 | 112 | |
More Information
Set Membership: |
|

TIMP4
Genomic Map