Assay Details
Gene Symbol: KIAA0753 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015296193.2 | XP_015151679.1 | 2 - 3 | 602 | 72 | |
XM_015296194.2 | XP_015151680.1 | 2 - 3 | 361 | 72 | ||
XM_015296198.2 | XP_015151684.1 | 2 - 3 | 331 | 72 | ||
XM_025142016.1 | XP_024997784.1 | 2 - 3 | 717 | 72 | ||
XM_025142017.1 | XP_024997785.1 | 3 - 4 | 545 | 72 | ||
XM_025142018.1 | XP_024997786.1 | 2 - 3 | 717 | 72 | ||
XM_025142019.1 | XP_024997787.1 | 2 - 3 | 717 | 72 | ||
XM_025142020.1 | XP_024997788.1 | 2 - 3 | 609 | 72 | ||
XM_025142021.1 | XP_024997789.1 | 3 - 4 | 430 | 72 | ||
XM_025142023.1 | XP_024997791.1 | 2 - 3 | 717 | 72 | ||
XM_025142024.1 | XP_024997792.1 | 2 - 3 | 717 | 72 | ||
GenBank mRNA | BX931537.1 | - | 2 - 3 | 725 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
