Assay Details
| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232592.5 | XP_001232593.3 | 3 | 7339 | 103 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRR16| Gene Symbol: PRR16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232592.5 | XP_001232593.3 | 3 | 7339 | 103 | |
Set Membership: |
|