Assay Details
| Gene Symbol: PI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004947027.3 | XP_004947084.1 | 2 - 3 | 253 | 91 | |
| GenBank mRNA | MG066611.1 | - | 2 - 3 | 297 | 91 | |
More Information
Set Membership: |
|

PI3
Genomic Map