Assay Details
| Gene Symbol: CDCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147502.1 | XP_025003270.1 | 8 | 4284 | 115 | |
| XM_418802.6 | XP_418802.3 | 9 | 4510 | 115 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CDCP1| Gene Symbol: CDCP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025147502.1 | XP_025003270.1 | 8 | 4284 | 115 | |
| XM_418802.6 | XP_418802.3 | 9 | 4510 | 115 | ||
Set Membership: |
|