Assay Details
| Gene Symbol: MKRN2OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414310.5 | XP_414310.2 | 3 - 4 | 614 | 136 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MKRN2OS| Gene Symbol: MKRN2OS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_414310.5 | XP_414310.2 | 3 - 4 | 614 | 136 | |
Set Membership: |
|