Assay Details
| Gene Symbol: TRIM62 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015297235.2 | XP_015152721.1 | 5 - 6 | 1160 | 102 | |
| XM_015297236.2 | XP_015152722.1 | 5 - 6 | 1035 | 102 | ||
More Information
Set Membership: |
|

TRIM62
Genomic Map