Assay Details
| Gene Symbol: CZH9ORF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025145254.1 | XP_025001022.1 | 14 - 15 | 2472 | 83 | |
| XM_025145255.1 | XP_025001023.1 | 14 - 15 | 2432 | 83 | ||
| XM_025145256.1 | XP_025001024.1 | 14 - 15 | 2444 | 83 | ||
| XM_025145257.1 | XP_025001025.1 | 12 - 13 | 2367 | 83 | ||
More Information
Set Membership: |
|

CZH9ORF3
Genomic Map