Assay Details
| Gene Symbol: ELFN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004937747.3 | XP_004937804.1 | 3 | 3574 | 101 | |
| XM_004937748.3 | XP_004937805.1 | 3 | 3183 | 101 | ||
| XM_015288333.2 | XP_015143819.1 | 2 | 3674 | 101 | ||
| XM_015288338.2 | XP_015143824.1 | 3 | 3178 | 101 | ||
| XM_015288343.2 | XP_015143829.1 | 3 | 3181 | 101 | ||
More Information
Set Membership: |
|

ELFN2
Genomic Map