Assay Details
| Gene Symbol: CHST13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233229.5 | XP_001233230.2 | 3 | 3845 | 60 | |
| XM_004944602.3 | XP_004944659.1 | 3 | 3886 | 60 | ||
More Information
Set Membership: |
|

CHST13
Genomic Map