Assay Details
| Gene Symbol: CHST12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015294348.2 | XP_015149834.1 | 2 | 1295 | 72 | |
| XM_015294349.2 | XP_015149835.1 | 2 | 1258 | 72 | ||
| XM_015294350.2 | XP_015149836.1 | 2 | 1574 | 72 | ||
| XM_015294351.2 | XP_015149837.1 | 2 | 2030 | 72 | ||
More Information
Set Membership: |
|

CHST12
Genomic Map