Assay Details
| Gene Symbol: PEBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025142908.1 | XP_024998676.1 | 4 - 5 | 460 | 78 | |
| GenBank mRNA | BX931846.1 | - | 4 - 5 | 461 | 78 | |
More Information
Set Membership: |
|

PEBP4
Genomic Map