Assay Details
| Gene Symbol: TTLL6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004943961.3 | XP_004944018.1 | 11 - 12 | 1648 | 67 | |
| XM_015292188.2 | XP_015147674.1 | 10 - 11 | 1504 | 67 | ||
| XM_015292189.2 | XP_015147675.1 | 10 - 11 | 2209 | 67 | ||
| XM_015292190.2 | XP_015147676.1 | 10 - 11 | 2331 | 67 | ||
| XM_015292191.2 | XP_015147677.1 | 11 - 12 | 2228 | 67 | ||
| XM_015292192.2 | XP_015147678.1 | 8 - 9 | 1192 | 67 | ||
| XM_025154050.1 | XP_025009818.1 | 11 - 12 | 1652 | 67 | ||
| XM_025154051.1 | XP_025009819.1 | 10 - 11 | 3491 | 67 | ||
More Information
Set Membership: |
|

TTLL6
Genomic Map