Assay Details
| Gene Symbol: SETBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233853.5 | XP_001233854.3 | 2 - 3 | 1154 | 110 | |
| GenBank mRNA | BX931980.1 | - | 1 - 2 | 241 | 110 | |
| BX934978.1 | - | 1 - 2 | 74 | 110 | ||
| BX935462.1 | - | 2 - 3 | 840 | 110 | ||
More Information
Set Membership: |
|

SETBP1
Genomic Map