Assay Details
| Gene Symbol: GUK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001232000.4 | XP_001232001.1 | 4 - 5 | 318 | 117 | |
| XM_003640644.3 | XP_003640692.1 | 3 - 4 | 226 | 117 | ||
| XM_425960.6 | XP_425960.2 | 3 - 4 | 217 | 117 | ||
| GenBank mRNA | BX932143.2 | - | 3 - 4 | 228 | 117 | |
| BX933011.1 | - | 3 - 4 | 192 | 117 | ||
| BX950363.2 | - | 3 - 4 | 226 | 117 | ||
| CR354011.1 | - | 3 - 4 | 225 | 117 | ||
| Gene Symbol: GJC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015281085.2 | XP_015136571.1 | 3 - 4 | 217 | 117 | |
| XM_015281086.2 | XP_015136572.1 | 3 - 4 | 217 | 117 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015281085.2, XM_015281086.2 transcripts.
Set Membership: |
|

GUK1
Genomic Map