Assay Details
| Gene Symbol: FFAR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003641481.4 | XP_003641529.3 | 1 - 2 | 1120 | 57 | |
| GenBank mRNA | JQ437358.1 | - | 1 - 2 | 533 | 57 | |
More Information
Set Membership: |
|

FFAR4
Genomic Map