Assay Details
| Gene Symbol: SMDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233648.5 | XP_001233649.1 | 1 - 2 | 219 | 70 | |
| GenBank mRNA | BX932653.2 | - | 1 - 2 | 179 | 70 | |
| BX932796.1 | - | 1 - 2 | 179 | 70 | ||
More Information
Set Membership: |
|

SMDT1
Genomic Map