Assay Details
| Gene Symbol: SMDT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233648.5 | XP_001233649.1 | 2 - 3 | 351 | 74 | |
| GenBank mRNA | BX932653.2 | - | 2 - 3 | 311 | 74 | |
| BX932796.1 | - | 2 - 3 | 311 | 74 | ||
| BX950530.2 | - | 2 - 3 | 319 | 74 | ||
More Information
Set Membership: |
|

SMDT1
Genomic Map