Assay Details
| Gene Symbol: SNRPD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_025144523.1 | XP_025000291.1 | 2 - 3 | 327 | 77 | |
| GenBank mRNA | BX933865.1 | - | 2 - 3 | 238 | 77 | |
| BX936185.1 | - | 2 - 3 | 257 | 77 | ||
More Information
Set Membership: |
|

SNRPD2
Genomic Map