Assay Details
| Gene Symbol: TIMM22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_415839.6 | XP_415839.5 | 3 - 4 | 705 | 68 | |
| GenBank mRNA | BX934001.2 | - | 3 - 4 | 507 | 68 | |
| BX934088.1 | - | 3 - 4 | 507 | 68 | ||
| BX935303.1 | - | 3 - 4 | 512 | 68 | ||
| CR353685.1 | - | 3 - 4 | 514 | 68 | ||
More Information
Set Membership: |
|

TIMM22
Genomic Map