Assay Details
| Gene Symbol: AGBL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001235270.5 | XP_001235271.4 | 2 - 3 | 163 | 200 | |
| XM_015291228.2 | XP_015146714.2 | 2 - 3 | 163 | 200 | ||
| XM_015291230.2 | XP_015146716.2 | 2 - 3 | 163 | 200 | ||
| XM_025153112.1 | XP_025008880.1 | 2 - 3 | 163 | 200 | ||
| GenBank mRNA | BX934320.2 | - | 2 - 3 | 166 | 200 | |
More Information
Set Membership: |
|

AGBL4
Genomic Map