Assay Details
| Gene Symbol: PRR15L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015299489.2 | XP_015154975.1 | 1 - 2 | 133 | 103 | |
| GenBank mRNA | BX934950.1 | - | 1 - 2 | 64 | 103 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015299489.2 transcripts.
Set Membership: |
|

PRR15L
Genomic Map