Assay Details
| Gene Symbol: GYG2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015277729.2 | XP_015133215.1 | 5 - 6 | 946 | 77 | |
| XM_015277730.2 | XP_015133216.1 | 5 - 6 | 946 | 77 | ||
| XM_015277731.2 | XP_015133217.1 | 5 - 6 | 946 | 77 | ||
| GenBank mRNA | BX935316.1 | - | 5 - 6 | 856 | 77 | |
More Information
Set Membership: |
|

GYG2
Genomic Map