Assay Details
| Gene Symbol: NDUFB7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015274582.2 | XP_015130068.1 | 2 - 3 | 329 | 92 | |
| GenBank mRNA | BX950780.2 | - | 2 - 3 | 305 | 92 | |
More Information
Set Membership: |
|

NDUFB7
Genomic Map