Assay Details
| Gene Symbol: RAB39B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420326.6 | XP_420326.4 | 2 | 8944 | 100 | |
| GenBank mRNA | CR352452.1 | - | 1 | 725 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAB39B| Gene Symbol: RAB39B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_420326.6 | XP_420326.4 | 2 | 8944 | 100 | |
| GenBank mRNA | CR352452.1 | - | 1 | 725 | 100 | |
Set Membership: |
|