Assay Details
| Gene Symbol: C3H1ORF198 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015284384.2 | XP_015139870.1 | 4 | 1939 | 99 | |
| GenBank mRNA | CR353600.1 | - | 2 | 1019 | 99 | |
More Information
Set Membership: |
|

C3H1ORF198
Genomic Map