Assay Details
| Gene Symbol: RASSF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015286726.2 | XP_015142212.1 | 5 | 76 | ||
| XM_015286727.2 | XP_015142213.1 | 4 | 76 | |||
| GenBank mRNA | CR386887.1 | - | 3 | 621 | 76 | |
More Information
Set Membership: |
|

RASSF7
Genomic Map