Assay Details
| Gene Symbol: FSBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233121.5 | XP_001233122.1 | 1 | 365 | 63 | |
| GenBank mRNA | CR391332.1 | - | 1 | 121 | 63 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

FSBP| Gene Symbol: FSBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001233121.5 | XP_001233122.1 | 1 | 365 | 63 | |
| GenBank mRNA | CR391332.1 | - | 1 | 121 | 63 | |
Set Membership: |
|