Assay Details
| Gene Symbol: TADA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426352.5 | XP_426352.1 | 2 | 1460 | 59 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TADA2B| Gene Symbol: TADA2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_426352.5 | XP_426352.1 | 2 | 1460 | 59 | |
Set Membership: |
|