Assay Details
| Gene Symbol: MAP3K8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282086.2 | XP_015137572.1 | 9 | 3076 | 64 | |
| XM_015282087.2 | XP_015137573.1 | 9 | 2202 | 64 | ||
| XM_025148361.1 | XP_025004129.1 | 8 | 2041 | 64 | ||
| XM_418581.6 | XP_418581.3 | 9 | 2221 | 64 | ||
More Information
Set Membership: |
|

MAP3K8
Genomic Map