Assay Details
| Gene Symbol: HS3ST3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295254.2 | XP_015150740.1 | 2 | 1240 | 152 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HS3ST3B1| Gene Symbol: HS3ST3B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015295254.2 | XP_015150740.1 | 2 | 1240 | 152 | |
Set Membership: |
|