Assay Details
| Gene Symbol: SERPINB12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015282196.2 | XP_015137682.1 | 4 - 5 | 952 | 73 | |
| XM_418985.5 | XP_418985.2 | 3 - 4 | 441 | 73 | ||
More Information
Set Membership: |
|

SERPINB12
Genomic Map