Assay Details
| Gene Symbol: LOC101748751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298099.2 | XP_015153585.1 | 5 - 6 | 1438 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101748751| Gene Symbol: LOC101748751 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015298099.2 | XP_015153585.1 | 5 - 6 | 1438 | 88 | |
Set Membership: |
|